Gold ≥99,999% (Metall-Basis), Drahtform, Premion®, Ø 2.0 mm (0.08 in)

Lieferant: Thermo Fisher Scientific

14733.BO 14733.BQ 14733.BU
14733.BOEA 174 EUR
14733.BO 14733.BQ 14733.BU
Gold ≥99,999% (Metall-Basis), Drahtform, Premion®, Ø 2.0 mm (0.08 in)
Gold

Purity: 99.999% CAS No.: 7440-57-5 Physical Form: ≈60.6g/m Odour: Odourless MDL No.: MFCD00003436

Formel: Au
Siedepunkt: 2800 °C (1013 hPa)
Schmelzpunkt: 1063 °C
Dichte: 19,3 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00003436
CAS-Nummer: 7440-57-5
EINECS: 231-165-9
Merck Index: 12,04526

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR