Detergent for use in special washers, liquid concentrate, neodisher® ProTech 5

Lieferant: Dr. Weigert
401433
148-0153EA 117 EUR
148-0153
Detergent for use in special washers, liquid concentrate, neodisher® ProTech 5
Reinigungsmittel Waschmittel und Oberflächenreiniger
Automated cleaning of printed circuit boards, plastic housing, assemblies, and other highly sensitive material in printed circuit board industry in the industrial sector.

  • Free of phosphates, oxidising agents, silicone compounds with good defoaming capacity
  • Viscosity: <100 mPas (concentrate, 20 °C)
  • Density: 1,0 g/cm³ (20 °C)
  • Sensitive to frost below –15 °C

Contains <30% non-ionic surfactants according to regulation (EC) No 648/2004 on detergents.

It is a special detergent in neutral pH formulation with excellent material compatibility. The special combination of special surfactants and solvents gives it high effective wetting and cleaning characteristics for the removal of fluxes and dust in low foam composition.

Bitte beachten For professional use only. Do not mix with other products.
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR