Detergents for glassware and plastics, Aquet®, SP Bel-Art
Lieferant: Bel-Art Products, a Part of SP
F170940030
148-1170EA 144 EUR
148-1170
Detergents for glassware and plastics, Aquet®, SP Bel-Art
Reinigungsmittel Waschmittel und Oberflächenreiniger
Phosphate-Free Liquid Detergent with Neutral pH
Safe for glassware, plastics and surfaces, this phosphate-free, nonionic surfactant will not etch or mar precision glassware. It rinses quickly and dries without wiping.