Regenerating salts for dishwashers
Lieferant: KAMMERER
1161340
148-2501EA 7.6 EUR
148-2501
Regenerating salts for dishwashers
Reinigungsmittel Waschmittel und Oberflächenreiniger
Regenerating salts for water softening systems for dishwashers, it protects dishes and machines from calcification. Also they ensures optimum efficiency of the dish cleaner and are meant for all cleaning systems.
  • Highly pure and residue-free soluble
  • Coarse-grained for optimal solubility
Standards: EN 973 Type A.