Alkaline detergents, powder, LaboClean PLM, neodisher®
Lieferant: Dr. Weigert
412276
148-3133EA 198 EUR
148-3133
Alkaline detergents, powder, LaboClean PLM, neodisher®
Reinigungsmittel Waschmittel und Oberflächenreiniger
Mildly alkaline detergent.
  • Free of oxidative additives
Suitable for the removal of the basic components of creams respective lotions and easily emulsified residues. With defoaming properties.