Alkaline detergents, powder, LaboClean F, neodisher®
Lieferant: Dr. Weigert
410581
148-3139EA 230 EUR
148-3139
Alkaline detergents, powder, LaboClean F, neodisher®
Reinigungsmittel Waschmittel und Oberflächenreiniger
LaboClean F detergent is composed of alkaline components and dispersants. Ideal for the removal of grease, oil, paraffin, resin, organic dyes and residues of indelible pen.
  • High complexing and dispersive action
  • Used for intensive cleaning
  • Free of phosphates and surfactants
It is not suitable for non ferrous and light metals.