Draining racks
Lieferant: WDF - Funke GmbH & Co. KG
5280-01-11
149-1350EA 197 EUR
149-1350
Draining racks
Gestelle Abtropfgestelle
Wire, PE-coated, white.
  • Very good chemical resistance
  • Very good temperature resistance
  • Easy to clean
It is designed with free-standing or for wall-mounting and with drip pan. The feet are protected by plastic caps.