Draining racks
Lieferant: WDF - Funke GmbH & Co. KG
5281-01-11
149-1351EA 408 EUR
149-1351
Draining racks
Gestelle Abtropfgestelle
Wire, PE-coated.
  • Very good chemical resistance
  • Very good temperature resistance
  • Easy to clean
It is designed with free-standing or for wall-mounting and with drip pan. The feet are protected by plastic caps.