1-Aminocyclopropancarbonsäure, Calbiochem®

Lieferant: Merck Millipore Calbiochem

Synonyme: 1-Amino-1-cyclopropanecarboxylic acid

MQ100

149101-1
149101-1EA 386 EUR
149101-1
1-Aminocyclopropancarbonsäure, Calbiochem®
1-Aminocyclopropanecarboxylic acid
Formel: C₄H₇NO₂
Molecular Weight: 101.11 g/mol
MDL: MFCD00009944
CAS-Nummer: 22059-21-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR