Bismarckbraun R für die Mikroskopie, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY

15000-100G
15000-100GEA 0 EUR
15000-100G
Bismarckbraun R für die Mikroskopie, Sigma-Aldrich®
Bismarckbraun R

Bismarck Brown R has been used as an adsorbate and is used in dye decoloration experiment.

Formel: C₂₁H₂₄N₈·2HCl
MDL: MFCD00036386
CAS-Nummer: 5421-66-9
EINECS: 226-541-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR