Isopentylnitrit (Isomerengemisch) 96%, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Gefahr

Synonyme: Isoamyl nitrite

M-Clarity™ Qualitätsstufe = MQ200

150495-100ML 150495-500ML 150495-5ML
150495-100MLEA 69.5 EUR
150495-100ML 150495-500ML 150495-5ML
Isopentylnitrit (Isomerengemisch) 96%, Sigma-Aldrich®
Isopentylnitrit (Isomerengemisch)

Isopentyl nitrite, also known as isoamyl nitrite, is an oxidant, and nitrosating agent for phosphoramidites which rearrange into phosphotriesters during nitrosation.


  • Isopentyl nitrite has been used as a radical initiator to functionalize multi-walled carbon nanotubes
  • As a reagent in the preparation of 4-carboxy-phenyl diazonium salt tetrafluoroborate
  • In the synthesis of peroxynitrite


Isopentyl nitrite, also known as isoamyl nitrite, is used for the generation of benzyne from anthranilic acid. It is a smooth muscle relaxant that may be suitable as nitrovasodilators and as recreational drugs known as poppers. Its applications are also seen in kits to treat cyanide poisoning, its function being to oxidise hemoglobin (Hb) to methemoglobin (metHb) which reacts with cyanide to form the non-toxic cyano-metHb. It is used a diagnostic assay manufacturing, hematology, histology.

Formel: C₅H₁₁NO₂
Molecular Weight: 117.15 g/mol
Siedepunkt: 99 °C
Schmelzpunkt: ≤80 °C
Dichte: 0,872 g/ml (25 °C)
MDL: MFCD00002057
CAS-Nummer: 110-46-3
EINECS: 203-770-8
UN: 1113
ADR: 3 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=07b0a40203d4f9eb2536f5e13063c9cd4bfae4bc&vid=0af94bf7cb703f39ac908a3f6db251ea5723e917&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance (Colour) Colorless to Yellow and Colourless to Green-Yellow
Appearance (Form) Liquid
Purity (GC AREA %) ≥95.5 %
Infrared spectrum Conforms to structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR