Barium silicon oxide

Lieferant: Thermo Scientific
Achtung

15122.36
15122.36EA 211 EUR
15122.36
Barium silicon oxide
Barium silicon oxide

CAS No.: 13255-26-0 Molecular Formula: BaO and SiO2 Formula Weight: 213.40 Physical Form: Powder Density: 4.399 Refractive Index: 1.67 Melting Point: 1604° UN No.: UN1564 MDL No.: MFCD00014186

Formel: BaO and SiO₂
Molecular Weight: 213,40 g/mol
Schmelzpunkt: 1604 °C
Dichte: 4,399
Lagertemperatur: Raumtemperatur
MDL: MFCD00014186
CAS-Nummer: 13255-26-0
UN: 1564
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR