Organic reference standard, Bisphenol A bis(3-chloro-2-hydroxypropyl) ether, analytical standard, Supelco®
Lieferant: Merck
15136-250MG
15136-250MGEA
132
EUR
15136-250MG
Organic reference standard, Bisphenol A bis(3-chloro-2-hydroxypropyl) ether, analytical standard, Supelco®
Standards
Organische Standards
Standard for determining toxic monomers released from polymers of the inner coating of cans.