Organic reference standard, Bisphenol A bis(3-chloro-2-hydroxypropyl) ether, analytical standard, Supelco®
Lieferant: Merck
15136-250MG
15136-250MGEA 132 EUR
15136-250MG
Organic reference standard, Bisphenol A bis(3-chloro-2-hydroxypropyl) ether, analytical standard, Supelco®
Standards Organische Standards
Standard for determining toxic monomers released from polymers of the inner coating of cans.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d149dfe182014e78e77b519f19cca995366fd80a&vid=687d89b7366d68aaffeb1a7a08fdada94915390c&prid=web&clid=SAPDAM