Organic reference standard, Bisphenol A bis(2,3-dihydroxypropyl) ether, analytical standard, Supelco®
Lieferant: Merck
15137-250MG
15137-250MGEA 239 EUR
15137-250MG
Organic reference standard, Bisphenol A bis(2,3-dihydroxypropyl) ether, analytical standard, Supelco®
Standards Organische Standards
Standard for determining toxic monomers released from polymers of the inner coating of cans.
Verpackung: Bottomless glass bottle. Contents are inside inserted fused cone.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=22f50f1d595fa6660b4603beadca000b1082adfa&vid=22f50f1d595fa6660b4603beadca000b1082adfa&prid=web&clid=SAPDAM