Organic reference standard, Bisphenol A diglycidyl ether, analytical standard, Supelco®
Lieferant: Merck
15138-500MG-F
15138-500MG-FEA 123 EUR
15138-500MG-F
Organic reference standard, Bisphenol A diglycidyl ether, analytical standard, Supelco®
Standards Organische Standards
Standard for determining toxic monomers released from polymers of the inner coating of cans.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6fd33f0cb096c2de6a507004393cdecbc163b3cd&vid=6fd33f0cb096c2de6a507004393cdecbc163b3cd&prid=web&clid=SAPDAM