Bis[4-(1,1,3,3-tetramethylbutyl)phenyl] phosphate calcium salt ≥97,0%, Selectophore™ Ionophor, Supelco®

Lieferant: Merck

Synonyme: hemi-Calcium bis[4-(1,1,3,3-tetramethylbutyl)phenyl] phosphate

M-Clarity™ Qualitätsstufe = MQ100

15180-1G
15180-1GEA 1500 EUR
15180-1G
Bis[4-(1,1,3,3-tetramethylbutyl)phenyl] phosphate calcium salt ≥97,0%, Selectophore™ Ionophor, Supelco®
Bis[4-(1,1,3,3-tetramethylbutyl)phenyl] phosphate calcium salt
Formel: C₅₆H₈₄CaO₈P₂
Molecular Weight: 987,29 g/mol
MDL: MFCD00043425
CAS-Nummer: 40835-97-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ceb0eb2c5704fc63f009256b0b5c5eb3c8ca635e&vid=33628d064bc25ba1e027e1669253542c2eb9985d&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR