Certriacetat Sesquihydrat ≥99,9% (REO, Basis der Oxide der Metalle der seltenen Erden)
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Essigsäure Cer(III)salz Sesquihydrat, Cer(III)acetat Sesquihydrat
With bromide ion, catalyzes the liquid-phase auto-oxidation of cresols.
Cerium(III) acetate sesquihydrate, acts as a source of cerium involved in the preparation of cerium oxide. It finds application in metallurgy, glass and glass polishing, ceramics, catalysts, and in phosphors. It is utilized in steel production to remove the free sulfur and oxygen. Further, it is an efficient glass polishing agent.
With bromide ion, catalyzes the liquid-phase auto-oxidation of cresols
Formel:
Ce(C₂H₃O₂)₃·1.5H₂O Molecular Weight: 344,28 g/mol (317,26 anhy) Schmelzpunkt: 308 °C (Zersetzung) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00051060 CAS-Nummer: 537-00-8 EINECS: 208-654-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...