Blei(II)oxid, yellow 99,0 Complexometric, Puriss. p.a. analytical reagent, Fluka™

Lieferant: Honeywell Chemicals
Gefahr

Synonyme: Bleimonoxid

15338-250G 15338-1KG 15338-25KG
15338-250GEA 198.72 EUR
15338-250G 15338-1KG HONC15338-25KG
Blei(II)oxid, yellow 99,0 Complexometric, Puriss. p.a. analytical reagent, Fluka™
Blei(II)oxid
Formel: PbO
Molecular Weight: 223.2 g/mol
Siedepunkt: 1.470 °C (1.013 hPa)
Schmelzpunkt: 890 °C
Dichte: 9.66 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011164
CAS-Nummer: 1317-36-8
EINECS: 215-267-0
UN: 2291
ADR: 6.1 III
Merck Index: 13,05431

Bestellen

Spezifikation Testergebnisse

Assay (complexometric) 99.0 - 101.0 %
Calcium (Ca) Max. 10 ppm
Cadmium (Cd) Max. 5 ppm
Cobalt (Co) Max. 5 ppm
Chromium (Cr) Max. 5 ppm
Copper (Cu) Max. 5 ppm
Iron (Fe) Max. 10 ppm
Potassium (K) Max. 50 ppm
Magnesium (Mg) Max. 5 ppm
Manganese (Mn) Max. 5 ppm
Sodium (Na) Max. 50 ppm
Nickel (Ni) Max. 5 ppm
Zinc (Zn) Max. 5 ppm
Chloride (Cl) Max. 20 ppm
Total N Max. 10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR