Aktivkohle, Pulver USP, J.T.Baker®
Lieferant: Avantor
Synonyme:
Carbon activated, Activated carbon, Activated charcoal
1560-01EA
0
EUR
1560-01
Aktivkohle, Pulver USP, J.T.Baker®
Aktivkohle
Formel:
C Molecular Weight: 12,01 g/mol Schmelzpunkt: 3550 °C Dichte: 2 g/cm³ (20 °C) |
MDL:
MFCD00133992 CAS-Nummer: 7440-44-0 EINECS: 231-153-3 UN: 1362 ADR: 4.2,III REACH: 01-2119488894-16 Merck Index: 13,01818 |
Spezifikation Testergebnisse
Meets U.S.P Requirements | |
GMP Manufactured Product | |
CAUTION: For Manufacturing, processing or repackaging | |
Bulk Pharmaceutical Chemical | Passes Test |
Microbial Limits - Escherichia coli | Passes Test |
Microbial Limits - Salmonella | Passes Test |
Reaction | Passes Test |
USP - Loss on Drying | ≤15.0 % |
Residue on Ignition | ≤4.0 % |
Acid-Soluble Substances | ≤3.5 % |
Chloride (Cl) | ≤0.2 % |
USP - Sulfate (SO₄) | ≤0.2 % |
USP - Sulfide (S) | Passes Test |
USP - Cyanogen Compounds | Passes Test |
USP - Uncarbonized Constituents | Passes Test |
Adsorptive Power: Alkaloids | Passes Test |
Adsorptive Power: Dyes | Passes Test |
Typical Particle Size, % thru 150 micron | ≥97 % |
No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...