Tetrachlorophthalonitrile ≥98%, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
|
Achtung
|
Synonyme:
3,4,5,6-Tetrachlorobenzene-1,2-dicarbonitrile, Perchlorophthalonitrile
158801-5G
158801-5GEA
260
EUR
158801-5G
Tetrachlorophthalonitrile ≥98%, Sigma-Aldrich®
Tetrachlorophthalonitrile
Tetrachlorophthalonitrile,98%
Formel:
C₈Cl₄N₂ Molecular Weight: 265,91 g/mol Schmelzpunkt: 249…252 °C (Zers., Lit.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00019740 CAS-Nummer: 1953-99-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...