VWR®, Gesamthärteindikator, Tabletten für die Bestimmung der Wasserhärte
Lieferant: VWR Chemicals
160232EEA 42.3 EUR
160232E
VWR®, Gesamthärteindikator, Tabletten für die Bestimmung der Wasserhärte
Umwelt-Prüfkits
Indicator tablets for determination of water hardness by titration with EDTA.
The tablets are dissolved in 100 ml of the water sample to be tested, which has been buffered at pH 10 with 1 ml of Ammonia/ Ammonium chloride buffer solution, and then titrated with EDTA solution (0,01 M) till the sample turns from red to blue.
Total hardness in mg/l = 10×ml of titrant used.
Control samples of calcium carbonate should be used to calibrate the method.