Tetrabutylammoniumhydroxid 55% (w/w) in wässriger Lösung
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
16198.18
16198.30
16198.A1
16198.18EA
67.7
EUR
16198.18
16198.30
16198.A1
Tetrabutylammoniumhydroxid 55% (w/w) in wässriger Lösung
Tetrabutylammoniumhydroxid
CAS No.: 2052-49-5
Molecular Formula: C16H37NO
Formula Weight: 259.48
Flash Point: None
Physical Form: Liquid
Density: 0.97
Refractive Index: 1.435
Sensitivity: Air Sensitive
Appearance: Colourless to light yellow
pH: 14
Beilstein Reference: 3571228
UN No.: 3267
MDL No.: MFCD00009425
As a reagent for ion-pair extractions
Formel:
C₁₆H₃₇NO Dichte: 0,97 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00009425 CAS-Nummer: 2052-49-5 EINECS: 218-147-6 UN: 3267 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...