Sulbactam sodium, Pulver
Lieferant: USP
|
Gefahr
|
Synonyme:
Sodium (2S-cis)-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate 4,4-dioxide
1623671
1623671.EA
344
EUR
1623671.
Sulbactam sodium, Pulver
Sulbactam sodium
Formel:
C₈H₁₀NO₅S·Na Molecular Weight: 255,223 g/mol |
MDL:
MFCD01750374 CAS-Nummer: 69388-84-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...