Thymolphthalein ACS
Lieferant: Thermo Fisher Scientific
Synonyme:
2-(1-Hydroxy-1,1-di(4-hydroxy-2-methyl-5-(methylethyl)phenyl)methyl)benzoesäurelacton, 3,3-Bis(4-hydroxy-5-isopropyl-o-tolyl)phthalid
16245.06
16245.14
16245.06EA
37.7
EUR
16245.06
16245.14
Thymolphthalein ACS
Thymolphthalein
CAS No.: 125-20-2
Molecular Formula: C28H30O4
Formula Weight: 430.55
Physical Form: Crystalline
Melting Point: 251-253°
Solubility: Insoluble in water. Soluble in alcohol, acetone, dilute alkalies (blue Colour), H2SO4 (red Colour)
Beilstein Reference: 359413
Merck Reference: 14,9401
MDL No.: MFCD00005909
pH indicator; Colourless below pH 9.3, blue above pH 10.5
Formel:
C₂₈H₃₀O₄ Schmelzpunkt: 251…253 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005909 CAS-Nummer: 125-20-2 EINECS: 204-729-7 Merck Index: 14,9401 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...