Gold(I)iodid ≥99%
Lieferant: Thermo Fisher Scientific
Purity: 99%
CAS No.: 10294-31-2
Molecular Formula: AuI
Formula Weight: 323.87
Physical Form: Crystalline
Density: 8.25
Melting Point: 120° dec.
Solubility: Insoluble in water. Soluble in alkali iodide, cyanide solutions. Decomposes in warm acids
UN No.: 3260
MDL No.: MFCD00014175
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...