Pinacyanol iodide 97%, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Gefahr

Synonyme: Iodide, Quinaldine Blue, 1,1'-Diethyl-2,2'-carbocyanine Iodide

M-Clarity™ Qualitätsstufe = MQ100

166510-1G 166510-250MG
166510-1GEA 410 EUR
166510-1G 166510-250MG
Pinacyanol iodide 97%, Sigma-Aldrich®
Pinacyanol iodide
Formel: C₂₅H₂₅IN₂
Molecular Weight: 480.39 g/mol
Schmelzpunkt: 295 °C (Zers., Lit.)
Lagertemperatur: Kühlschrank
MDL: MFCD00011975
CAS-Nummer: 605-91-4
EINECS: 210-099-4
ADR: 6.1 II

Bestellen

Spezifikation Testergebnisse

Appearance Dark green to black powder, crystals or Crystalline powder
Infrared spectrum Conforms to structure.
Uv-visible spectrum In ethanol
Carbon 60.3%-64.7%
Nitrogen 5.5%- 6.1%
Thin-layer Consistent with 97% purity
Solubility 10 mg/ml(1%), ethanol; dark blue solution


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR