Bis-(4-bromphenyl)-ether ≥99%
Lieferant: Thermo Fisher Scientific
16684.06
16684.14
16684.06EA
36.1
EUR
16684.06
16684.14
Bis-(4-bromphenyl)-ether ≥99%
Bis(4-bromophenyl)ether
Purity: 99%
CAS No.: 2050-47-7
Molecular Formula: C12H8Br2O
Formula Weight: 328.01
Physical Form: Crystalline
Appearance: White
Melting Point: 56-60°
Boiling Point: 338-340°
MDL No.: MFCD00000095
Formel:
C₁₂H₈Br₂O Siedepunkt: 338…340 °C Schmelzpunkt: 56…60 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000095 CAS-Nummer: 2050-47-7 EINECS: 218-090-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...