Cäsiumiodid 99,9995 Metals basis, TraceSELECT™, Fluka™

Lieferant: Honeywell Chemicals
Achtung

Synonyme: Cesiumiodide

16722-25G
16722-25GEA 189.33 EUR
16722-25G
Cäsiumiodid 99,9995 Metals basis, TraceSELECT™, Fluka™
Cäsiumiodid

TraceSELECT™ Ultra and TraceSELECT


  • Ultra-pure acids, bases, and salts for smelting and wet digestion in environmental, water, and food analysis
  • Sample preparation for trace analysis requires reagents of the highest purity; our TraceSELECT Ultra products for ultra-trace analysis at ppb and even ppt levels are produced by sub-boiling distillation
  • The acids, bases, and salts in the TraceSELECT series have been developed for sample preparation and analysis in the ppb (µg/kg) trace range
  • packaged in high-quality containers appropriate for each product
  • the reagents are produced and bottled under clean-room conditions

Formel: CsI
Molecular Weight: 259.81 g/mol
Siedepunkt: 1280
Schmelzpunkt: 621
Lagertemperatur: Raumtemperatur
MDL: MFCD00010962
CAS-Nummer: 7789-17-5
EINECS: 232-145-2
Merck Index: 12,02056

Bestellen

Spezifikation Testergebnisse

Aluminium (Al) Max. 0.02 ppm
Arsenic (As) Max. 0.05 ppm
Barium (Ba) Max. 0.1 ppm
Beryllium (Be) Max. 0.005 ppm
Calcium (Ca) Max. 0.1 ppm
Cadmium (Cd) Max. 0.02 ppm
Cerium (Ce) Max. 0.01 ppm
Cobalt (Co) Max. 0.005 ppm
Chromium (Cr) Max. 0.01 ppm
Copper (Cu) Max. 0.005 ppm
Dysprosium (Dy) Max. 0.01 ppm
Erbium (Er) Max. 0.01 ppm
Europium (Eu) Max. 0.01 ppm
Iron (Fe) Max. 0.05 ppm
Gallium (Ga) Max. 0.005 ppm
Gadolinium (Gd) Max. 0.01 ppm
Mercury (Hg) Max. 10 ppb
Holmium (Ho) Max. 0.01 ppm
Indium (In) Max. 0.005 ppm
Iridium (Ir) Max. 0.5 ppm
Potassium (K) Max. 0.5 ppm
Lanthanum (La) Max. 0.01 ppm
Lithium (Li) Max. 0.1 ppm
Lutetium (Lu) Max. 0.01 ppm
Magnesium (Mg) Max. 0.05 ppm
Manganese (Mn) Max. 0.01 ppm
Molybdenum (Mo) Max. 0.01 ppm
Sodium (Na) Max. 0.2 ppm
Neodymium (nd) Max. 0.01 ppm
Nickel (Ni) Max. 0.01 ppm
Lead (Pb) Max. 0.01 ppm
Palladium (Pd) Max. 0.5 ppm
Praseodymium (Pr) Max. 0.01 ppm
Platinum (Pt) Max. 0.5 ppm
Rubidium (Rb) Max. 0.1 ppm
Rhodium (Rh) Max. 0.5 ppm
Ruthenium (Ru) Max. 0.5 ppm
Antimony (Sb) Max. 0.05 ppm
Scandium (Sc) Max. 0.01 ppm
Samarium (Sm) Max. 0.01 ppm
Strontium (Sr) Max. 0.05 ppm
Terbium (Tb) Max. 0.01 ppm
Thorium (Th) Max. 0.01 ppm
Titanium (Ti) Max. 0.01 ppm
Thulium (Tm) Max. 0.01 ppm
Uran (U) Max. 0.01 ppm
Vanadium (V) Max. 0.01 ppm
Yttrium (Y) Max. 0.01 ppm
Ytterbium (Yb) Max. 0.01 ppm
Zinc (Zn) Max. 0.1 ppm
Total S (as SO₄) Max. 10 ppm
Bromide, chloride (as Cl) Max. 100 ppm
Phosphate (PO₄) Max. 10 ppm
Iodate (IO₃) Max. 2 ppm
Total N Max. 10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR