Azomethin H Hydrat

Lieferant: Thermo Fisher Scientific

16810.04 16810.09 16810.14
16810.04EA 72.1 EUR
16810.04 16810.09 16810.14
Azomethin H Hydrat
Azomethin H Hydrat

CAS No.: 32266-60-7 Molecular Formula: C17H13NO8S2.xH2O Formula Weight: 423.40(anhy) Physical Form: Crystalline MDL No.: MFCD00066536

Formel: C₁₇H₁₃NO₈S₂·H₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD00066536
CAS-Nummer: 32266-60-7
EINECS: 250-975-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR