Cobalt(II,III)oxid, Pulver, BAKER ANALYZED®, Laborreagens, J.T.Baker®
Lieferant: Avantor
Synonyme:
Trikobalttetraoxid, Tricobalttetraoxid
1688-09EA
0
EUR
1688-09
Cobalt(II,III)oxid, Pulver, BAKER ANALYZED®, Laborreagens, J.T.Baker®
Cobalt(II,III)oxid
Formel:
Co₃O₄ Molecular Weight: 240,8 g/mol |
MDL:
MFCD00010939 CAS-Nummer: 1308-06-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...