Cobalt(II,III)oxid, BAKER ANALYZED®, J.T.Baker®

Lieferant: Avantor

1691-08EA 0 EUR
1691-08 1691-09
Cobalt(II,III)oxid, BAKER ANALYZED®, J.T.Baker®
Cobalt(II,III)oxid
Formel: Co₃O₄
MDL: MFCD00010939
CAS-Nummer: 1308-06-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR