Sepharose™ 4B Gel Filtration Base Matrix
Lieferant: Cytiva
17-0120-01
17-0120-01EA
1020
EUR
17-0120-01
Sepharose™ 4B Gel Filtration Base Matrix
Protein-Aufbereitungs
Protein Purification Resins
Sepharose™ 4B is an agarose gel filtration base matrix and is frequently used for coupling affinity ligands to the matrix. The matrix is not pre-activated and the user performs all steps in coupling.
- 4% agarose gel filtration media
- Proven base matrix for coupling affinity ligands