Sepharose™ CL-6B Gel Filtration Base Matrix
Lieferant: Cytiva
17-0160-01
17-0160-01EA
1090
EUR
17-0160-01
Sepharose™ CL-6B Gel Filtration Base Matrix
Protein-Aufbereitungs
Protein Purification Resins
Sepharose™ CL-6B is a cross-linked agarose gel filtration base matrix and is frequently used for coupling affinity ligands to the matrix.
- 6% cross-linked agarose gel filtration media
- Proven base matrix for coupling affinity ligands
- Higher degree of crosslinking improve matrix stability
The matrix is not pre-activated and the user performs all steps in coupling.