Cell culture reagents, microcarriers, Cytodex 1
Lieferant: Cytiva
17-0448-01 17-0448-02 17-0448-03
17-0448-01EA 456 EUR
17-0448-01 17-0448-02 17-0448-03
Cell culture reagents, microcarriers, Cytodex 1
Kugeln Mikropartikel
Cytodex 1 is particularly suitable for most established cell lines and for production of viruses or cell products from cultures of primary cells and normal diploid cell strains.
  • General-purpose microcarrier for most established, anchorage-dependent cell lines
  • Contains positive charges throughout the microporous matrix
  • Transparent for easy microscopic examination of attached cells
Bestellen
TECHNISCHE DATEN
Environmentally Preferable