Agarose IEF
Lieferant: Cytiva
17-0468-01
17-0468-01EA
297
EUR
17-0468-01
Agarose IEF
Agarose
Agarose with low electroendosmosis (EEO) is recommended for most DNA/RNA applications.
Agarose IEF is suitable for isoelectric focusing of large molecules and is the gel of choice for many IEF applications and immunochemical detection methods.
If separated nucleic acids are to be used as substrates for enzymes (e.g., ligase and restriction endonucleases), a special highly purified agarose such as Agarose NA should be used.
Schmelzpunkt:
60…90 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00081294 CAS-Nummer: 9012-36-6 EINECS: 232-731-8 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...