Superose™ 12 Gel Filtration Media, Prep Grade
Lieferant: Cytiva
17-0536-01
17-0536-01EA 760 EUR
17-0536-01
Superose™ 12 Gel Filtration Media, Prep Grade
Protein-Aufbereitungs Protein Purification Resins
Good resolution separations of proteins and other biomolecules according to size, Wide separation range – separations in two molecular weight ranges from 1,000 up to 5 million, Excellent reproducibility and durability
  • Excellent reproducibility and durability
Achieve high-resolution separations across exceptionally broad molecular-weight ranges. Low ionic strength eluents (< 0.05 M) can utilize a hydrophobic interaction component to alter the selectivity of Superose for some lipids, peptides, and small aromatic compounds. High performance gel filtration media specially designed for preparative purification of biomolecules. Superose 6 is designed for high-resolution chromatography. Superose 6 prep grade permits easy, efficient scale-up to preparative separations. Superose 12 is for high resolution purification of proteins from 1 × 103 to 3 × 105 molecular weight, while Superose 12 prep grade is useful for preparative purification of these higher molecular weight proteins.