Hydrophobic interaction chromatography media, Phenyl Sepharose™ High Performance
Lieferant: Cytiva
17-1082-01
17-1082-01EA 484 EUR
17-1082-01
Hydrophobic interaction chromatography media, Phenyl Sepharose™ High Performance
Protein-Aufbereitungs Protein Purification Resins
Phenyl Sepharose™ High Performance is an aromatic hydrophobic interaction chromatography (HIC) medium, designed for intermediate and polishing step purification steps when high resolution has priority.
  • Selectivity similar to Phenyl Sepharose™ 6 Fast Flow (low sub) but higher resolution due to the smaller 34 µm bead size
  • Bioprocess medium supported for industrial applications and well-established in approved processes
  • The hydrophilic nature of the base matrix ensures low levels of non-specific binding