Affinitätschromatographie-Kit, MAbTrap™
Lieferant: Cytiva
17-1128-01
17-1128-01EA 518 EUR
17-1128-01
Affinitätschromatographie-Kit, MAbTrap™
Chromatographiesäulen
MAbTrap™ is an affinity chromatography kit for fast and effective purification of monoclonal and polyclonal IgG from ascites fluid, serum and cell culture supernatants. MAbTrap™ Kit contains everything necessary to perform a complete one step purification without using complicated equipment. It also eliminates time consuming buffer preparation and column packing.
  • Purification of IgG from monoclonal and polyclonal material using a syringe in <15 minutes
Lieferumfang: Contains one HiTrap™ Protein G HP column prepacked with Protein G Sepharose™ High Performance and enough buffer concentrate for 20 preparations with a syringe.