Natriumdodecylsulfat (SDS), PlusOne™

Lieferant: Cytiva
Gefahr

17-1313-01
17-1313-01EA 100.35 EUR
17-1313-01
Natriumdodecylsulfat (SDS), PlusOne™
Natriumdodecylsulfat (SDS)

SDS is an ionic detergent used to solubilise proteins. The PlusOne™ SDS reagent is carefully controlled for carbon chain length and overall quality.


  • pH Range: 6,5 - 8,5
  • Partially soluble in cold water, hot water and methanol,
  • Very slightly soluble in acetone, insoluble in diethyl ether

Formel: CH₃(CH₂)₁₁OSO₃Na
Schmelzpunkt: 204…207 °C
Dichte: 1,1 g/cm³ (20 °C)
Flammpunkt: >100 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00036175
CAS-Nummer: 151-21-3
EINECS: 205-788-1
UN: 8027
ADR: 4.1,III
Merck Index: 14,08636
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe4cb1741fba41f699a3be006d0b01b1b551898c&vid=c6df9a86fc351b319e68cc79c1c17f733b4fec67&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR