Glycin (H-Gly-OH), PlusOne™

Lieferant: Cytiva

Synonyme: H-Gly-OH, Glycocoll, Aminoessigsäure

17-1323-01
17-1323-01EA 86.88 EUR
17-1323-01
Glycin (H-Gly-OH), PlusOne™
Glycin (H-Gly-OH)

Glycine is frequently used as a component of electrophoresis buffers, especially in the Laemmli discontinuous buffer system for SDS-PAGE.


  • Soluble in cold water and hot water
  • Slightly soluble in acetone, insoluble in diethyl ether

Formel: NH₂CH₂COOH
Molecular Weight: 75.07 g/mol
Siedepunkt: 181
Schmelzpunkt: 245 °C
Dichte: 1.16 g/cm³
Lagertemperatur: Raumtemperatur
MDL: MFCD00008131
CAS-Nummer: 56-40-6
EINECS: 200-272-2
Merck Index: 13,04504
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d944007d30d5d4650062014a1b152bd9bfb66ea3&vid=093f4227cbdd1c4c6515ef9c4b85d93799877dcc&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR