Affinity chromatography media, Benzamidine Sepharose™ 4 Fast Flow (High Sub)
Lieferant: Cytiva
17-5123-10
17-5123-10EA 473 EUR
17-5123-10
Affinity chromatography media, Benzamidine Sepharose™ 4 Fast Flow (High Sub)
Protein-Aufbereitungs Protein Purification Resins
Benzamidine Sepharose™ 4 Fast Flow (High Sub) is a tool for removal and/or purification of trypsin and trypsin-like serine proteases, as well as zymogens, including urokinase and prekallikrein.
  • For affinity chromatography
  • p-aminobenzamidine (pABA) immobilized on Sepharose 4 Fast Flow matrix gives high productivity, and allows easy scale-up
  • Effective removal of thrombin and factor Xa after tag cleavage of recombinant proteins
  • Amide coupling method
  • Stable in all commonly used aqueous buffers
Removal of serine proteases is easily done directly from serum, monoclonal cell supernatants and bacterial lysates. Benzamidine Sepharose™ 4 Fast Flow (High Sub) is based on highly cross-linked 4% agarose enabling high flow rates without losing binding capacity.
For convenient, one-step removal and/or purification of trypsin, trypsin-like serine proteases, and zymogens including urokinase and prekallikrein.