Affinity chromatography columns, HiTrap™ Benzamidine FF
Lieferant: Cytiva
17-5143-01 17-5143-02 17-5144-01
17-5143-01EA 436 EUR
17-5143-01 17-5143-02 17-5144-01
Affinity chromatography columns, HiTrap™ Benzamidine FF
Chromatographiesäulen
HiTrap Benzamidine FF (high sub) are prepacked with Benzamidine Sepharose 4 Fast Flow (high sub) and designed specifically for the removal and/or purification of serine proteases such as thrombin.
  • Effective removal of thrombin and factor Xa after tag cleavage of recombinant proteins
  • Removal of serine proteases is easily done directly from serum, monoclonal cell supernatants and bacterial lysates
  • High binding capacity
  • It can be operated using syringe, peristaltic pump or liquid chromatography system such as ÄKTA design or FPLC system
HiTrap Benzamidine FF (high sub) are prepacked 1 ml and 5 ml columns for convenient, one-step removal and/or purification of trypsin, trypsin-like serine proteases, and zymogens including urokinase and prekallikrein.
The medium, Benzamidine Sepharose 4 Fast Flow (high sub), is also available as lab packages and is an excellent choice for scale-up.