HiTrap columns, Phenyl FF (Low Sub)
Lieferant: Cytiva
17-5193-01 17-5194-01
17-5193-01EA 520 EUR
17-5193-01 17-5194-01
HiTrap columns, Phenyl FF (Low Sub)
Chromatographiesäulen
Phenyl Sepharose 6 Fast Flow is a well-established, standard aromatic HIC medium for capture and intermediate purification requiring low to medium hydrophobicity.
  • Provide convenient and fast small-scale protein purifications.
  • Phenyl Sepharose 6 Fast Flow is a well-established, standard aromatic HIC medium for capture and intermediate purification requiring low to medium hydrophobicity. Two levels of substitution on Phenyl Sepharose 6 Fast Flow help find optimal selectivity and binding capacity.
  • Flexible design for use with syringes, single laboratory pump, or chromatography system such as ÄKTA design.
  • High flow rates and high capacities.
  • Excellent and easy scale-up achieved by connecting several HiTrap columns in series or using HiPrep columns.
Two levels of substitution on Phenyl Sepharose 6 Fast Flow help find optimal selectivity and binding capacity. HiTrap Phenyl FF (Low Sub) are prepacked Phenyl Sepharose 6 Fast Flow columns for hydrophobic interaction chromatography (HIC). The columns are used for small-scale protein purification as well as for screening of binding and elution conditions.