Affinity chromatography media, HiTrap™ Capto™ Lentil Lectin
Lieferant: Cytiva
17-5489-11
17-5489-11EA 1382 EUR
17-5489-11
Affinity chromatography media, HiTrap™ Capto™ Lentil Lectin
Protein-Aufbereitungs Protein Purification Resins
Capto™ Lentil Lectin is an affinity chromatography resin for purification of glycoproteins and other molecules containing carbohydrates such as α-D-mannose and α-D-glucose or sterically related residues.
  • Group-specific adsorbent for carbohydrate-containing molecules
  • High productivity and cost-efficiency in downstream operations
  • Animal component-free production
  • Security of supply and regulatory support
Capto™ Lentil Lectin resin is based on a rigid high-flow agarose base matrix that enables high flow rates. The product is available in bulk as well as in various high-throughput process development formats.