Affinity chromatography media, MabSelect™ PrismA
Lieferant: Cytiva
17-5498-01 17-5498-02
17-5498-01EA 2515 EUR
17-5498-01 17-5498-02
Affinity chromatography media, MabSelect™ PrismA
Protein-Aufbereitungs Protein Purification Resins
MabSelect™ PrismA has been improved with an optimised high-flow agarose base matrix and a genetically engineered protein A-derived ligand, allowing future demands in mAb processing to be met.
  • Enhanced dynamic binding capacity allows high mass throughput of processed mAb per resin volume unit
  • Excellent alkaline stability enables efficient cleaning and sanitisation using 0,5 to 1,0 M NaOH for improved process economy, bioburden, control and robustness
  • Covered by a comprehensive security of supply program, including dual sources of the agarose base matrix and protein A ligand
MabSelect™ PrismA builds on the proven track record of MabSelect™ and MabSelect™ SuRe™ resins in commercial mAb production.