Water Guard, arium®
Lieferant: Sartorius
610AWG1
171-0899EA 506 EUR
171-0899
Water Guard, arium®
Lecksuchsysteme Flüssigkeits-Lecksuchsysteme
Arium® Water guard detects water leaks early. That means the lab gets optimal protection against water damage.
  • High-grade material, non-corrosive
  • Optic and acoustic alarm
  • Easy to install
  • Integrated wall holder for solenoid valve
  • Versatile
Leaks are registered by an optical sensor. This automatically closes the inlet line and triggers an acoustic warning signal. In addition, the user can also constantly monitor the system's status on the built-in LED display.