Zirkonium(IV)iodid ≥99%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
17400.14
17400.14EA
606
EUR
17400.14
Zirkonium(IV)iodid ≥99%
Zirkonium(IV)iodid
Purity: 99%
CAS No.: 13986-26-0
Molecular Formula: I4Zr
Formula Weight: 598.84
Physical Form: Powder
Density: 4.85
Appearance: Orange
Melting Point: 431° subl.
Merck Reference: 14,10178
UN No.: 3260
MDL No.: MFCD00049641
Formel:
I₄Zr Schmelzpunkt: 431 °C (Subl.) Dichte: 4,85 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00049641 CAS-Nummer: 13986-26-0 EINECS: 237-780-9 UN: 3260 ADR: 8,III Merck Index: 14,10178 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...