Blei(II)trifluoracetat-Halbhydrat

Lieferant: Thermo Scientific
Gefahr

17553.09
17553.09EA 121 EUR
17553.09
Blei(II)trifluoracetat-Halbhydrat
Blei(II)trifluoracetat-Halbhydrat

It is employed in pyrolytic deposition of fluoride glass.


CAS No.: 4146-73-0 Molecular Formula: C4F6O4Pb.0.5H2O Formula Weight: 442.23(433.22anhy) Physical Form: Powder Appearance: White Odour: Odourless UN No.: 2291 MDL No.: MFCD00061594


Bitte beachten Store in a cool dry place in tightly closed containers. Incompatible with oxidizing agents. Air and moisture sensitive.

Formel: C₄F₆O₄Pb·0.5H₂O
Schmelzpunkt: 147…153 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00061594
CAS-Nummer: 4146-73-0
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR