tetra-Natriumorthosilicat (Mixture of NaOH and Na2SiO3 yielding ≈ Na4SiO4 in solution)

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Natriumorthosilicat, Tetranatriumorthosilicat

17568.A1
17568.A1EA 60.4 EUR
17568.A1
tetra-Natriumorthosilicat (Mixture of NaOH and Na2SiO3 yielding ≈ Na4SiO4 in solution)
tetra-Natriumorthosilicat

It is used as a degreasant, industrial alkaline detergents, water treatment (boiler-feed); adhesives, builder in soaps.


CAS No.: 13472-30-5 Molecular Formula: Na4SiO4 Physical Form: Crystalline Appearance: White Odour: Odourless UN No.: 1759 MDL No.: MFCD00014252

Formel: Na₄O₄Si
Molecular Weight: 184.04 g/mol
Schmelzpunkt: 1078
Dichte: 2.5 g/cm³ (26 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00014252
CAS-Nummer: 13472-30-5
EINECS: 236-741-3
UN: 1759
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR