tri-Natriumcitrat Dihydrat, BAKER, J.T.Baker®

Lieferant: Avantor

Synonyme: Natriumcitrat tribasisch Dihydrat, Citronensäure Trinatriumsalz Dihydrat

1758.1000EA 57.11 EUR
1758.1000
tri-Natriumcitrat Dihydrat, BAKER, J.T.Baker®
tri-Natriumcitrat Dihydrat
Formel: Na₃C₆H₅O₇·2H₂O
Molecular Weight: 294.1 g/mol
Schmelzpunkt: 150
Dichte: 1.76 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00150031
CAS-Nummer: 6132-04-3
EINECS: 200-675-3
Merck Index: 13,08675
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e32147a215e8895f4b6db5aab1b99da667b795ac&vid=b929cda9d6f2eed18a8fefe57aa02b561af7c97f&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Meets Reagent Specifications for testing Ph. Eur. monographs.
Assay (anhydrous substance) 99.0 - 100.5 %
Acidity or Alkalinity passes test
Appearance of solution passes test
Chloride (Cl) max. 0.003 %
Heavy Metals (as Pb) max. 10 ppm
Identification passes test
Oxalates (C2O4) max. 300 ppm
Readily Carbonisable Substances passes test
Sulfate (SO4) max. 0.005 %
Water (H2O) 11.0 - 13.0 %
Additional Specification(s):
Tartrate passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR